Learn More
Medchemexpress LLC Necrocide 1 | 1247028-61-0 | 98.8% | C23H27NO3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1430710MG
Encompass_Preferred
Necrocide 1 is a small-molecule research compound described as a selective agonist of the TRPM4 channel (CAS 1247028-61-0). Supplied as a white to off-white solid and as a DMSO solution, it is intended for in vitro pharmacology and mechanistic studies of TRPM4-mediated pathways.
  • Selective TRPM4 agonist for in vitro studies.
  • Available as solid and DMSO solution for flexible dosing.
  • High purity to support reproducible bioassays.
  • Compact molecular weight suitable for cell permeability studies.
  • Provided with chemical formula and structural identifiers for cheminformatics.
Catalog No. 50-002-61337