Learn More
Medchemexpress LLC NecroIr1 | 98.0% | 823.36 g/mol | C40H29ClIrN5O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14836510MG
Encompass_Preferred
NecroIr1 is an iridium(III) coordination complex used as a research reagent to induce necroptosis in cisplatin-resistant lung cancer cells (A549R). It preferentially accumulates in mitochondria, promotes reactive oxygen species (ROS) generation and loss of mitochondrial membrane potential, activates necroptosis-associated kinases, and induces G0/G1 cell-cycle arrest that inhibits cell proliferation in vitro.
  • High purity (98.02%) suitable for cellular assays.
  • Preferential mitochondrial accumulation enabling targeted ROS generation.
  • Activates necroptosis signaling through RIPK1, RIPK3, and MLKL.
  • Induces G0/G1 cell-cycle arrest and inhibits proliferation in resistant lung cancer cells.
  • Provided as a solid 10 mg unit for laboratory studies.
  • Store sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-87396