Learn More
Medchemexpress LLC NecroIr1 | 98.5% | 823.36 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14836510MM/1ML
Encompass_Preferred
NecroIr1 is an iridium(III) complex that acts as a necroptosis inducer in Cisplatin-resistant lung cancer cells (A549R). It accumulates selectively in mitochondria, causing oxidative stress and a decrease in mitochondrial membrane potential (MMP). NecroIr1 activates receptor-interacting serine-threonine kinase 3 (RIPK3) and Mixed Lineage Kinase (MLKL), and regulates CDK4 expression.
  • Accumulates over 90% in mitochondria
  • Increases ROS generation and causes a loss of mitochondrial membrane potential
  • Activates necroptosis proteins and increases phosphorylation of RIPK1 and RIPK3
  • Induces necroptosis by arresting cell cycle at G0/G1
  • Inhibits A549R cell proliferation
Catalog No. 50-003-62175