Learn More
TARGETMOL CHEMICALS INC NECROPTOSIS-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T93055MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Necroptosis-IN-1 is a potent necroptosis inhibitor of RIPK1. It is an analog of Necrostatin-1. purity: 99%
Catalog No. 50-257-9009