Learn More
Enzo Life Sciences Necrostatin-1, (20mg), CAS Number: 4311-88-0
SDP
Supplier:  Enzo Life Sciences BMLAP3090020
Encompass_Preferred
Inhibits necroptosis. Necrostatin-1 inhibits necroptosis, a non-apoptotic cell death pathway. Inhibits the loss of mitochondrial membrane potential in TNFα-treated Jurkat cells (EC.50=180nM). Alternative Name: Methylthiohydantoin-DL-tryptophan. Formula: C13H13N3OS. MW: 259.3. CAS Number: 4311-88-0. Purity: ≥98% (TLC). Appearance: Yellow solid. Solubility: Soluble in DMSO (20mg/ml) or 100% ethanol (5mg/ml). Use/Stability: Stock solutions are stable for up to 3 months when stored at -20°C. Long Term Storage: Ambient
Catalog No. NC0918510