Learn More
Medchemexpress LLC Necrostatin-34 | 375835-43-1 | 98.9% | 384.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY132203
Encompass_Preferred
Necrostatin-34 is a RIPK1 kinase inhibitor that stabilizes RIPK1 kinase in an inactive conformation by occupying a distinct binding pocket in the kinase domain. It is intended for research use only.
  • RIPK1 kinase inhibitor
  • Stabilizes RIPK1 kinase in an inactive conformation
  • Occupies a distinct binding pocket in the kinase domain
  • Inhibits dimerization-induced RIPK1 activation
  • May block TNFα-induced complex II formation
Catalog No. 50-003-64275