Learn More
Cayman Chemical Necrostatin5 Inhibitors
SDP
Supplier:  Cayman Chemical 1052725 MG
Encompass_Preferred
A RIP1 kinase inhibitor; inhibits phosphorylation of RIP1 in Jurkat cells at 10 and 30 µM; inhibits TNF-α-induced necroptosis in FADD-deficient Jurkat cells (EC50 = 0.24 µM); reduces LDH release in S. marcescens-infected MH-S macrophages; improves survival in a mouse model of S. marcescens-induced pneumonia at 100 µM, i.p.
Catalog No. 50-289-0333