Learn More
Medchemexpress LLC Necrosulfonamide-d4 | 1795144-22-7 | 99.3% | 465.50 | C18H11D4N5O6S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY100573S5MG
Encompass_Preferred
Necrosulfonamide-d4 is the deuterium-labeled analogue of necrosulfonamide, a small-molecule inhibitor of MLKL and gasdermin D used in studies of necroptosis and pyroptosis. It is intended for research use as an isotope-labeled reference standard and tracer in biochemical and mass-spectrometry applications.
  • Deuterium-labeled analogue for tracer and quantitative MS studies.
  • Molecular weight 465.50 g/mol.
  • High purity (≈99.3%) suitable for analytical applications.
  • Acts as an inhibitor of MLKL and gasdermin D for cell death pathway research.
  • Supplied in pre-weighed, small-quantity vials for convenient handling.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-11779