Learn More
BPS Bioscience Inc Nectin-4 Lentivirus, Size: 500 µl x 2
SDP
Supplier:  BPS Bioscience Inc 78712
Encompass
The Nectin-4 Lentiviruses are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to transduce nearly all types of mammalian cells, including primary and non-dividing cells. The particles contain a human Nectin-4 (NM_ 030916.2) driven by an EF1A promoter and a puromycin selection marker (Figure 1).
Catalog No. NC2748344