Learn More
Selleck Chemical LLC Nedometinib-E1902-100MG
SDP
Supplier:  Selleck Chemical LLC E1902100MG
Encompass_Preferred
Nedometinib (NFX-179) is a highly specific and potent inhibitor of MEK1 with an IC50 of 135 nM It also results in suppression of p-ERK levels and can be used in research and treatment of cutaneous neurofibromas (cNFs) associated with neurofibromatosis type 1 (NF1)
Catalog No. 50-313-7801