Learn More
Med Vet International Terumo Needle, 23GX 1in, Hypodermic, 100/BX, NN2325R
Supplier:  Med Vet International 2325RN
Encompass_Preferred
This Terumo NN2325R Needle comes with a 23GX 1' Thin Wall needle. Terumo's thin wall needles areprecisely honed and beveled. This makes for ease of penetration and less patient discomfort This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-252-2570
 Restricted Item