Learn More
Med Vet International BD Needle, 25G x 5/8in, Hypodermic, 100/Box, 305122
Supplier:  Med Vet International 305122
Encompass_Preferred
This BD 305122 Needle is a 25G X 5/8' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-252-2553