Learn More
Trajan Scientific Needle Diamond MS Bevel 1/EA
SDP
Supplier:  Trajan Scientific 0355332
Encompass_Preferred
Ultra smooth 50 mm 0 50 mm OD 0 20 mm ID bevel tipped needle for 100-500 L manual MS syringes
Catalog No. 50-004-94974