Learn More
World Precision Instrument Halsey Needle Holder, 14cm, Straight, Serrated
SDP
Supplier:  World Precision Instrument 14110
Encompass
Halsey needle holders are used to hold and manipulate surgical needles during procedures. Halsey needle holders have a scissor-like jaw design with a flat, blunt tip. They have a locking mechanism to secure the jaws as they grasp the needle firmly. While Halsey needle holders are available in sizes from 4 to 7 inches, these are 5.5" long. Choose the size of your needle holders (also called needle drivers) based on the size of your needles. 
Catalog No. 50-822-213