Learn More
World Precision Instrument Crile-Wood Needle Holder, 14.5cm, Serrated, TC Inserts
SDP
Catalog No. 50822395
Encompass
Crile Wood needle holders are widely used in medical procedures. Named after Dr. George Crile, Sr. of the Cleveland Clinic and English ophthalmologist Dr. Charles Richard Wood who invented them, the Crile Wood needle holders are a standard surgical instrument for delicate surgeries. They are designed with a ratcheted locking mechanism, allowing surgeons to securely grasp and hold needles in place while suturing. The ratchet mechanism lets you securely locking the instrument in a closed position to firmly grip the needle, providing stability and control during suturing. The tips and jaws of Crile Wood needle holders are delicate and finely serrated, designed for a firm grasp without damaging the needle. These Crile Wood needle holders have tungsten carbide inserts incorporated into the jaws. Tungsten carbide is a very hard material, which enhances the instrument's ability to grip and hold the needle firmly without slipping.
Catalog No. 50-822-395 Supplier World Precision Instrument Supplier No. 500224
Crile Wood needle holders are widely used in medical procedures. Named after Dr. George Crile, Sr. of the Cleveland Clinic and English ophthalmologist Dr. Charles Richard Wood who invented them, the Crile Wood needle holders are a standard surgical instrument for delicate surgeries. They are designed with a ratcheted locking mechanism, allowing surgeons to securely grasp and hold needles in place while suturing. The ratchet mechanism lets you securely locking the instrument in a closed position to firmly grip the needle, providing stability and control during suturing. The tips and jaws of Crile Wood needle holders are delicate and finely serrated, designed for a firm grasp without damaging the needle. These Crile Wood needle holders have tungsten carbide inserts incorporated into the jaws. Tungsten carbide is a very hard material, which enhances the instrument's ability to grip and hold the needle firmly without slipping.