Learn More
Med Vet International Miltex Halsey Needle Holder, Serrated, Tungsten, 4000 Teeth P/Sq.in, 5.25in, German, 8-8A-Tc
Supplier:  Med Vet International MIL88ATC
Encompass_Preferred
Halsey needle holders are most commonly used for holding suture needles during surgical procedures. It features a cross-hatched and reliable non-slip surface that allows firm gripping of needles during surgical procedures. Ideally used for plastic surgery. Put together with medical grade German steel. Serrated jaws for improved grip. 5.25in total length. 4000 teeth per square " Tungsten carbide inlays provide increased durability against corrosion and abrasions.
Catalog No. 50-283-2328