Learn More
Med Vet International Miltex Swedish Needle Holder, Serrated Jaws, Tungsten, 5.87in, German, Pm-2426
Supplier:  Med Vet International MILPM2426
Encompass_Preferred
Sarot needle holders are designed to grip smaller needles. The shank is curved inward close to the finger rings while they are extended outward. Tungsten carbide inlays provide increased durability and precision. 5.87in total length. Handcrafted with medical grade German steel.
Catalog No. 50-282-9133