Learn More
Med Vet International Derf Needle Holder, 4.75-in, Economy
Supplier:  Med Vet International 111200
Encompass_Preferred
The Derf needle holder features short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures. Frequently utilized in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf needle holder to be easily controlled in smaller surgical areas.
Catalog No. 50-222-4039