Learn More
Med Vet International Dynarex Hypodermic Needle, Non-Safety, 23g, 1" Needle, 1000/Case
Supplier:  Med Vet International 6970
Encompass_Preferred
The Dynarex Hypodermic Needles are designed to provide a comfortable, effective way to deliver medication or draw blood. Color-coded needle hub for quick identification of the needle gauge Compatible with luer lock and luer slip syringes Non-Toxic Non-Pyrogenic Not made with natural rubber latex
Catalog No. 50-283-0228