Learn More
Med Vet International Jorvet Osty Core Bone Biopsy Needle, 16g x 3in, J0116C, Each
Supplier: Med Vet International JOR116C
The Jorvet osty core bone biopsy needle has a diameter of 16g and a length of 3in. This needle is for taking core samples of bone, and should NOT be used for bone marrow aspirates. This product is sterile and not autoclavable. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.