Learn More
Med Vet International FlipLock Safety Needles, 27G x 1in, 50/Box
Supplier:  Med Vet International 812701
Encompass_Preferred
Perhaps the most versatile and cost-effective safety needle available. EasyTouch FlipLock safety needles fit all standard Luer Lock and Slip Fit syringe barrels. Easy to Use Protective Cover Shields Needle After Use Multi-lock Safety System Latex Free
Catalog No. 50-264-3616