Learn More
Thermo Scientific Needle Seat 1 Ea
Supplier:  Thermo Scientific 6252.2470
Encompass
The needle seat of the Vanquish Neo autosampler represents the 1500 bar pressure capable interface between the sampler needle unit and the autosampler switching valve. The needle seat contains a 0.5 μm filter frit, which prevents insoluble components, which may have been drawn up into the needle from entering the system.
Catalog No. NC2093824