Learn More
Med Vet International Terumo Needle, 22GX 1in, Hypodermic, 100/BX, NN2225R
Supplier:  Med Vet International 2225RN
Encompass_Preferred
This Terumo NN2225R Needle comes with a 22GX 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This quantity is 100/BX
Catalog No. 50-209-2545
 Restricted Item