Learn More
SEKISUI DIAGNOSTICS LLC NEFA-HR(2) SOLVENT B
Supplier:  SEKISUI DIAGNOSTICS LLC 99335191
Encompass_Preferred
The HR Series NEFA-HR (2) is an in vitro enzymatic colorimetric method assay for the quantitative determination on non-esterified fatty acids (NEFA) in serum.
Catalog No. NC9567464