Learn More
Medchemexpress LLC Nefiracetam | 77191-36-7 | 99.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB03401G
Encompass_Preferred
Nefiracetam is a cognition-enhancing agent that activates nAChR, N-methyl-D-aspartate receptor (NMDAR), mGluR5, PKC, gamma-aminobutyric acid (GABA), and N/L-type Ca2+ channels. It promotes neuroplasticity and enhances neuroprotection. This compound is used in research related to Alzheimer's disease, epilepsy, and cerebral ischemia.
  • Activates nAChR, NMDAR, mGluR5, PKC, GABA, and N/L-type Ca2+ channels
  • Promotes neuroplasticity and neuroprotection
  • Supports research in Alzheimer's disease, epilepsy, and cerebral ischemia
  • Increases hippocampal neurotransmission
  • Improves NMDA receptor-dependent long-term potentiation
  • Enhances CaMKIIα and PKCα autophosphorylation
Catalog No. 50-004-06198