Learn More
Medchemexpress LLC HY-10328 10mg Medchemexpress, Neflamapimod CAS:209410-46-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1032810MG
Encompass_Preferred
Medchemexpress, HY-10328 10mg Neflamapimod CAS:209410-46-8 Neflamapimod (VX-745) is a potent, blood-brain barrier penetrant, highly selective inhibitor of p38α inhibitor with an IC50 for p38α of 10 nM and for p38β of 220 nM. Neflamapimod (VX-745) possesses anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9530