Learn More
Medchemexpress LLC Nefopam (hydrochloride) | 23327-57-3 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1057500MG
Encompass_Preferred
Nefopam hydrochloride is a centrally-acting, non-opioid analgesic drug used for the relief of moderate to severe pain. It also targets β-catenin protein levels in mesenchymal cells both in-vitro and in-vivo.
  • Functions as a pain reliever for moderate to severe pain.
  • Is a non-opioid drug.
  • Targets β-catenin protein levels in mesenchymal cells.
Catalog No. 50-004-13755