Learn More
Biotium Negative Control for Rabbit Monoclonal Antibodies (NCRBM/1520R), CF405S conjugate, 0.1mg/mL
Supplier:  Biotium BNC041520500
Encompass_Preferred
To serve as a true negative control for various procedures, this rabbit monoclonal IgG should be used under the identical conditions and at the same dilution as the rabbit monoclonal antibody being used for a positive reaction. Available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability.
Catalog No. 50-008-08923