Learn More
TARGETMOL CHEMICALS INC NEGLETEIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2S08435MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Negletein (7-O-Methylbaicalein) has anti-inflammatory and immunomodulatory activities via inhibition of TNF-(alpha) iNOS and IL-1(beta). 2. Negletein as a neuroprotectant enhances the action of nerve growth factor and induces neurite outgrowth in PC12 cells. 3. Negletein shows a comparable redox-active potential and it (50 uM 4 h) can activate Nrf2. 4. Negletein shows significant antimicrobial activity. 5. Negletein has anti-hypoxia activity it can significantly prolong the survival time of hypoxic mice.6. Negletein has anti- Alzheimer's disease activity it inhibits the iron-dependent formation of ROS and also blocks the iron-induced oligomerization of amyloid beta 42 in vitro. 7. Negletein (IC50 3.89 +/- 0.39 microM) exhibits potent inhibitory activity against nuclear factor of activated T cells (NFAT) transcription factor. purity: 99%
Catalog No. 50-258-0438