Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NIMA-related kinase 2 (NEK2) protein, human | >80.0% | 50UG
SDP

Supplier:  Medchemexpress LLC HYP70273950UG

Encompass_Preferred

Recombinant active human NEK2 kinase fragment (residues M1-I271) expressed in E. coli and supplied as a filtered solution formulated for stability. Intended for biochemical kinase assays, inhibitor screening, and research applications that require active NEK2 enzymatic activity.

  • Active human NEK2 fragment (M1-I271).
  • Produced in E. coli for recombinant expression.
  • Approximate molecular weight 32.3 kDa.
  • Supplied as a 0.22 μm filtered solution in 50 mM HEPES, pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • Purity >80% by reducing SDS-PAGE.
  • Tag free to avoid interference in assays.
  • Available in small unit sizes suitable for assay development.
  • Store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-001-75148


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.