Learn More
Bon Opus Biosciences Recombinant Human NELF-E(C-His) - Source: E. coli
SDP
Supplier:  Bon Opus Biosciences BP287650UG
Encompass_Preferred
Recombinant Human NELF-E is produced by E.coli expression system and the target gene encoding I4-F380 is expressed with a His tag at the C-terminus. Accession#P18615. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733764