Learn More
Medchemexpress LLC Nelfinavir | 159989-64-7 | C32H45N3O4S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15287
Encompass_Preferred
Nelfinavir is a potent and orally bioavailable inhibitor of HIV-1 protease, exhibiting a Ki of 2 nM, and is utilized for HIV infection. It also acts as a broad-spectrum anticancer agent.
  • Potent and orally bioavailable protease inhibitor.
  • Inhibits protease with a Ki of 2 nM.
  • Broad-spectrum anticancer activity.
  • Treatment of HIV infection.
  • Anticancer research.
  • Use as an analytical standard.
  • Isotope-labeled for specific research.
Catalog No. 50-003-74706