Learn More
Apexbio Technology LLC Nelfinavir 159989-64-7 10mM (in 1mL Ethanol)
SDP
Supplier:  Apexbio Technology LLC B11225.1
Encompass_Preferred
Nelfinavir is a selective inhibitor of HIV-1 protease which is responsible for cleaving viral precursor proteins into functional units essential for HIV replication By disrupting this proteolytic step nelfinavir inhibits viral maturation and subsequent infectivity In biochemical assays nelfinavir demonstrates inhibitory activity against HIV-1 protease with an IC50 value typically ranging from 2 to 10 nM Due to its defined mechanism of action nelfinavir is commonly utilized in biomedical research focusing on HIV pathogenesis including antiviral activity studies viral resistance profiling and therapeutic combination assessments in vitro and in cell-based experimental models
Catalog No. 50-000-62131