Learn More
Medchemexpress LLC HY-15287 5mg Medchemexpress, Nelfinavir CAS:159989-64-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY152875MG
Encompass_Preferred
Medchemexpress, HY-15287 5mg Nelfinavir CAS:159989-64-7 Nelfinavir (AG-1341) is a potent and orally bioavailable HIV-1 protease inhibitor ( K i =2 nM) and antiviral agent for the treatment of HIV infection. Nelfinavir is a broad-spectrum, anticancer agent [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2731