Learn More
Medchemexpress LLC Nelfinavir (Mesylate) | 159989-65-8 | 98.9% | 663.89 | 5 MG
Supplier: Medchemexpress LLC 5MGHY15287A
Nelfinavir Mesylate is a potent and orally bioavailable HIV-1 protease inhibitor (Ki=2 nM) for HIV infection. It also acts as a broad-spectrum, anticancer agent. This compound inhibits the proliferation of multiple myeloma cells, induces apoptosis, and decreases AKT phosphorylation. It has also been identified as a SARS-CoV 3CLpro inhibitor.
- Potent and orally bioavailable HIV-1 protease inhibitor
- Broad-spectrum anticancer agent
- Inhibits proliferation of multiple myeloma cells
- Induces apoptosis of multiple myeloma cell lines
- Decreases AKT phosphorylation
- Acts as a SARS-CoV 3CLpro inhibitor
- Decreases multiple myeloma cell growth in vivo