Learn More
Medchemexpress LLC Nelonemdaz potassium | 916214-57-8 | 99.9% | 421.31 g/mol | C15H7F7KNO3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY106408A10MM 1ML
Encompass_Preferred
Nelonemdaz potassium is an NR2B-selective, noncompetitive N-methyl-D-aspartate (NMDA) receptor antagonist and free radical scavenger used in neuroprotection research. Supplied as a research-grade potassium salt, it is suitable for in vitro and preclinical studies and is available as DMSO solutions and solid powder.
  • Selective NR2B NMDA receptor antagonist.
  • Free radical scavenger with reported neuroprotective effects.
  • Available as 10 mM solution in DMSO and multiple solid quantities.
  • High purity appropriate for research applications.
  • Commonly used in studies of ischemia and NMDA receptor pharmacology.
Catalog No. 50-002-42084