Learn More
Medchemexpress LLC Nelonicline (ABT-126) | 1026134-63-3 | 99.8% | 313.42 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY167481MG
Encompass_Preferred
Nelonicline (ABT-126) is an orally active and selective α7 nicotinic receptor agonist. It has high affinity for α7 nAChRs in the human brain (Ki=12.3 nM) and is used in the research of schizophrenia and Alzheimer's disease. It activates currents in Xenopus oocytes expressing recombinant human α7 nAChRs (EC50=2 μM; intrinsic activity of 74% relative to acetylcholine) and binds to α3β4* nAChRs in human IMR-32 neuroblastoma cells (Ki=60 nM). Nelonicline is also a 5-HT3 receptor antagonist.
  • Orally active and selective α7 nicotinic receptor agonist
  • High affinity to α7 nAChRs in human brain (Ki=12.3 nM)
  • Used for the research of schizophrenia and Alzheimer's disease
  • Activates currents in Xenopus oocytes expressing recombinant human α7 nAChRs (EC50=2 μM)
  • Binds to α3β4* nAChRs in human IMR-32 neuroblastoma cells (Ki=60 nM)
  • 5-HT3 receptor antagonist
Catalog No. 50-003-90720