Learn More
Medchemexpress LLC Nelonicline (ABT-126) | 1026134-63-3 | 99.8% | 313.42 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1674850MG
Encompass_Preferred
Nelonicline (ABT-126) is an orally active and selective α7 nicotinic receptor agonist with high affinity to α7 nAChRs in human brain. It is used for the research of schizophrenia and Alzheimer's disease.
  • Selective α7 nicotinic receptor agonist
  • High affinity to α7 nAChRs
  • Used for research of schizophrenia and Alzheimer's disease
  • Binds to α3β4* nAChRs with lower efficacy
  • 5-HT3 receptor antagonist with lower affinity
  • Orally active
Catalog No. 50-003-93528