Learn More
Medchemexpress LLC Nemiralisib | 1254036-71-9 | 99.8% | 440.54 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19535A
Encompass_Preferred
Nemiralisib (GSK2269557 free base) is a potent and highly selective PI3Kδ inhibitor. It shows over 1000-fold selectivity over related PI3K isoforms, and inhibits IFNγ in PBMC assays.
  • Potent and highly selective PI3Kδ inhibitor
  • Active in disease-relevant animal models of lung inflammation
  • Suitable for research use
Catalog No. 50-004-03385