Learn More
Medchemexpress LLC Nemorubicin | 108852-90-0 | MFCD00871079 | 643.6 g/mol | C32H37NO13 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1579410MG
Encompass_Preferred
Nemorubicin is an anthracycline derivative of doxorubicin with potent antitumor activity; it intercalates into DNA and can stabilize G-quadruplex structures, and its cytotoxic action has been reported to require an intact nucleotide excision repair (NER) system.
  • Anthracycline derivative with potent cytotoxic activity.
  • Active against a variety of tumor cell lines, including multidrug-resistant phenotypes.
  • Intercalates into duplex DNA and stabilizes G-quadruplex structures.
  • Mechanism of action associated with dependence on nucleotide excision repair.
  • Available in multiple forms, including hydrochloride and GMP-grade variants.
Catalog No. 50-002-88339