Learn More
Electron Microscopy Sciences NEO Micro Coverglass, 24 x 40mm, size: 24 x 50mm., 100/PK
SDP
Supplier:  Electron Microscopy Sciences 7220702
Encompass_Preferred
This Special high-quality flat glass has a workable memory that makes it flexible enough to withstand the stress of automated coverslippers. A special coating privent slips from sticking to each other or reaching to environmental conditions, with no optical interference. Suitable for long-term storage. It is extremely stress resistant and environmentally friendly. size: 24 x 50mm.
Catalog No. 50-209-1008