Learn More
Medchemexpress LLC Neo Spiramycin I-d3 | 70253-62-2 | C36H59D3N2O11 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY143623S500UG
Encompass_Preferred
Neo Spiramycin I-d3 is the deuterium-labeled form of Neo Spiramycin I, an active metabolite of Spiramycin that exhibits antimicrobial activity. It is widely used in veterinary medicine.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are integrated into drug molecules as tracers for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-55551