Learn More
Selleck Chemical LLC Neoandrographolide S9157-1MG
SDP
Supplier:  Selleck Chemical LLC S91571MG
Encompass_Preferred
Neoandrographolide a principle diterpenoids isolated from A paniculata exerts a wide range of therapeutic actions including immunosuppressant antithrombotic anti-inflammatory antineoplastic anti-viral anti-bacterial anti-diabetic anti-oxidative stress antipyretic anti-edematogenic and anti-nociceptive activities
Catalog No. 50-000-25225