Learn More
TARGETMOL CHEMICALS INC NEOANDROGRAPHOLIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T38845MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Neoandrographolide (Neoandrographiside) has potent hypolipidemic effect and protects the cardiovascular without significant liver damage.Neoandrographolide has anti-inflammatory effect might result from inhibition of iNOS and COX-2 expression through inhibiting p38 MAPKs activation.Neoandrographolide as chemosensitizer in S-Jurkat and X chromosome-linked inhibitor of apoptosis protein (XIAP)-overexpressing Jurkat cells a model for chemoresistance. purity: 99%
Catalog No. 50-258-0082