Learn More
Medchemexpress LLC Neochebulagic acid | 28196-46-5 | 10 MG

Supplier: Medchemexpress LLC HYN690910MG
Neochebulagic acid is isolated from Phyllanthus emblica L., a plant from the Combretaceae family. This compound is classified under phenols and polyphenols. It presents as a white to off-white solid with a molecular weight of 972.68 and a chemical formula of C41H32O28. It is intended for research use only.
- Isolated from Phyllanthus emblica L.
- Molecular weight of 972.68
- Chemical formula: C41H32O28
- Appears as a white to off-white solid
- Classified under phenols and polyphenols
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.