Learn More
Medchemexpress LLC HY-119720 5mg Medchemexpress, Neocryptotanshinone CAS:109664-02-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1197205MG
Encompass_Preferred
Medchemexpress, HY-119720 5mg Neocryptotanshinone CAS:109664-02-0 Neocryptotanshinone, a fatty diterpenoids from Salvia Miltiorrhiza, inhibits lipopolysaccharide-induced inflammation by suppression of NF-κB and iNOS signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0931