Learn More
Medchemexpress LLC Neocuproine | 484-11-7 | 208.26 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00456310G
Encompass_Preferred
Neocuproine is an organic compound commonly used as a complexing reagent and copper ion detector. It forms stable complexes with copper ions and can act catalytically in chemical reactions and analytical methods. It is also utilized in biomedical fields, such as in the study of copper metabolism disorders and neurodegenerative diseases.
  • Used as a complexing reagent and copper ion detector
  • Forms stable complexes with copper ions
  • Acts catalytically in chemical reactions and analytical methods
  • Utilized in biomedical fields for studying copper metabolism disorders and neurodegenerative diseases
Catalog No. 50-004-48773