Learn More
TARGETMOL CHEMICALS INC NEODIOSMIN 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2P2898100MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Neodiosmin (Diosmetin-7-O-neohesperidoside) has poor aqueous solubility but exerts a good debittering effect and is a strong antioxidant with potential applications in foods beverages and pharmaceutical preparations. purity: 99%
Catalog No. 50-258-0440