Learn More
Medchemexpress LLC Neodiosmin | 38665-01-9 | 5 MG

Supplier: Medchemexpress LLC HYN41225MG
Neodiosmin is a flavone glycoside isolated from the leaves of Citrus aurantium. It is for research use only.
- Molecular weight: 608.54
- Formula: C28H32O15
- Appearance: Solid, light yellow to yellow
- Classified as a flavonoid, flavone, phenol, and polyphenol
- Solubility in DMSO (in vitro): 250 mg/mL
- In vivo solubility protocol available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.