Learn More
Medchemexpress LLC Neodiosmin | 38665-01-9 | 50 MG

Supplier: Medchemexpress LLC HYN412250MG
Neodiosmin is a flavone glycoside isolated from the leaves of Citrus aurantium.
- Purity: 99.37%
- Molecular weight: 608.54
- Formula: C₂₈H₃₂O₁₅
- Appearance: Solid, light yellow to yellow
- Initial source: Plants (Rutaceae, Citrus x aurantium Linnaeus)
- Classification: Natural product, flavonoid, phenol, polyphenol
- Related to disease research fields such as infection and inflammation/immunology
- Acts as an inhibitor
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.